Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Tetraethylammonium bromide 99%

Supplier: Ambeed
Warning
Synonyms: Tetrylammonium bromide

A1458816-25G A1458816-1000G A1458816-100G A1458816-500G
76661-870EA 65.92 USD InStock
76661-870 76661-872 76661-874 76661-876
Tetraethylammonium bromide 99%
Tetraethylammonium bromide
Caution: for Research Use Only


Formula: C₈H₂₀BrN
MW: 210.16 g/mol
MDL Number: MFCD00011825
CAS Number: 71-91-0
Order Now
Specification Test Results
Loss on drying ≤ 2%

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty