Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

5-Aminofluorescein (isomer I) 95%

Supplier: Ambeed
Warning
Synonyms: 5-Amino-3',6'-dihydroxyspiro[isobenzofuran-1(3H),9'-(9H)xanthen]-3-oneFluoresceinamine, isomer I5-Amino-3',6'-dihydroxy-3H-spiro[isobenzofuran-1,9'-xanthen]-3-one

A1467346-250MG A1467346-5G A1467346-1G
76662-558EA 151.41 USD InStock
76662-558 76662-556 76662-554
5-Aminofluorescein (isomer I) 95%
5-Aminofluorescein (isomer I)
Caution: for Research Use Only


Formula: C₂₀H₁₃NO₅
MW: 347.33 g/mol
MDL Number: MFCD00005052
CAS Number: 3326-34-9
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty