Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1-Bromoheptane 99%

Supplier: Ambeed
Danger
Synonyms: 1-Heptyl bromide

A1471056-25G
76662-754EA 6.18 USD InStock
76662-754
1-Bromoheptane 99%
1-Bromoheptane
Caution: for Research Use Only


Formula: BrH₂C(CH₂)₅CH₃
MW: 179.1 g/mol
MDL Number: MFCD00000273
CAS Number: 629-04-9
UN: 1993
ADR: 3,Ⅲ
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty