Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

(1R,2R)-(-)-1,2-Diaminocyclohexane ≥ 98% (99.9%ee)

Supplier: Ambeed
Danger
Synonyms: (1R,2R)-Cyclohexane-1,2-diamine(R,R)-DACH

A1471231-25G A1471231-5G
76662-800EA 10.3 USD InStock
76662-800 76662-798
(1R,2R)-(-)-1,2-Diaminocyclohexane ≥ 98% (99.9%ee)
(1R,2R)-(-)-1,2-Diaminocyclohexane
Caution: for Research Use Only


Formula: C₆H₁₄N₂
MW: 114.19 g/mol
MDL Number: MFCD00062985
CAS Number: 20439-47-8
UN: 2735
ADR: 8,Ⅲ
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty