Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Bromo-2-nitrotoluene 98%

Supplier: Ambeed
Warning

A1481650-5G A1481650-25G A1481650-500G
76664-168EA 207.03 USD InStock
76664-168 76664-166 76664-170
4-Bromo-2-nitrotoluene 98%
4-Bromo-2-nitrotoluene
Caution: for Research Use Only


Formula: C₇H₆BrNO₂
MW: 216.03 g/mol
MDL Number: MFCD00041243
CAS Number: 60956-26-5
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty