Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Amino-2-fluorobenzoic acid 98%

Supplier: Ambeed
Danger

A1485706-100G A1485706-5G A1485706-25G A1485706-1G
76664-854EA 6.18 USD InStock
76664-854 76664-852 76664-858 76664-856
4-Amino-2-fluorobenzoic acid 98%
4-Amino-2-fluorobenzoic acid
Caution: for Research Use Only


Formula: C₇H₆FNO₂
MW: 155.13 g/mol
MDL Number: MFCD01569397
CAS Number: 446-31-1
UN: 1759
ADR: 8,Ⅲ
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty