Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Aluminium acetate basic hydrate 98%

Supplier: Ambeed
Danger
Synonyms: Diacetoxy(hydroxy)aluminum

A1147190-100G A1147190-500G
76673-518EA 14.42 USD InStock
76673-518 76673-520
Aluminium acetate basic hydrate 98%
Aluminium acetate basic hydrate
Caution: for Research Use Only


Formula: C₄H₇AlO₅·H₂O
MW: 162.07 g/mol
MDL Number: MFCD00008688
CAS Number: 142-03-0
UN: 3077
ADR: 9,Ⅲ
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty