Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sodium chloride 99.9%, Analytical reagent (AR)

Supplier: Ambeed
Warning
Synonyms: SaltHaliteSea saltRock saltSaline

A1165388-500G
76675-790EA 6.18 USD InStock
76675-790
Sodium chloride 99.9%, Analytical reagent (AR)
Sodium chloride
Caution: for Research Use Only


Formula: NaCl
MW: 58.44 g/mol
MDL Number: MFCD00003477
CAS Number: 7647-14-5
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty