Synonyms:
Dihydrogen oxide
901-004-1X6
901-004-1X6
76683-186CS
102.02
USD
InStock
CA76683-186
76683-186
Water, GemPure™, Cell and Molecular Grade
Water
GeminiBio’s Cell and Molecular Grade Water is manufactured specifically for your most stringent research applications. GemPure™ Cell and Molecular Grade Water is sterile and low endotoxin for your cell culture processes. Protease and nuclease free, it is perfect for any of your molecular biology needs as well saving valuable lab space and simplifying your purchasing process.
- Tested for RNase, DNase, Protease, sterility, TOC, endotoxin, and more
- Manufactured in an ISO 13485:2016 and 21CFR820 compliant facility
- Available in 1 L bottle and custom sizes to suit your needs
- For further manufacturing or research use only
- Not for direct therapeutic use