Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Benzo[d]isoxazol-3-ol ≥96%

Supplier: APOLLO SCIENTIFIC
Warning
Synonyms: 3-Hydroxy-1,2-benzisoxazole

OR27158-1G OR27158-10G OR27158-5G
76571-994EA 189 USD InStock
76571-994 76571-992 76571-990
Benzo[d]isoxazol-3-ol ≥96%
Benzo[d]isoxazol-3-ol


Formula: C₇H₅NO₂
MW: 135.12 g/mol
Melting Pt: 137-140 °C
MDL Number: MFCD00125030
CAS Number: 21725-69-9
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty