Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3,5-Di-tert-butyl-4-hydroxybenzyl alcohol ≥95%

Supplier: APOLLO SCIENTIFIC
Danger

OR2794-5G OR2794-1G OR2794-25G
76572-324EA 53 USD InStock
76572-324 76572-320 76572-322
3,5-Di-tert-butyl-4-hydroxybenzyl alcohol ≥95%
3,5-Di-tert-butyl-4-hydroxybenzyl alcohol


Formula: C₁₅H₂₄O₂
MW: 236.35 g/mol
Boiling Pt: 138-141 °C
Melting Pt: 214 °C
MDL Number: MFCD00017254
CAS Number: 88-26-6
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty