Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

(S)-1-Boc-2-isopropylpiperazine

Supplier: APOLLO SCIENTIFIC
Warning
Synonyms: (S)-1-Boc-2-Isopropylpiperazine(2S)-2-Isopropylpiperazine, N1-BOC protected

OR15647-100MG
76567-870EA 13 USD InStock
76567-870
(S)-1-Boc-2-isopropylpiperazine
(S)-1-Boc-2-isopropylpiperazine


Formula: C₁₂H₂₄N₂O₂
MW: 228.34 g/mol
MDL Number: MFCD03787918
CAS Number: 674792-05-3
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty