Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Cyclopentyltriphenylphosphonium bromide ≥98%

Supplier: APOLLO SCIENTIFIC
Warning

OR6823-50G OR6823-250G OR6823-10G
76587-148EA 63 USD InStock
76587-148 76587-150 76587-152
Cyclopentyltriphenylphosphonium bromide ≥98%
Cyclopentyltriphenylphosphonium bromide


Formula: C₂₃H₂₄BrP
Melting Pt: 260-262 °C
MDL Number: MFCD00051880
CAS Number: 7333-52-0
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty