Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

5-Bromo-2-cyclopropylbenzoic acid ≥95%

Supplier: APOLLO SCIENTIFIC

OR70324-250MG OR70324-1G
76587-610EA 189 USD InStock
76587-610 76587-612
5-Bromo-2-cyclopropylbenzoic acid ≥95%
5-Bromo-2-cyclopropylbenzoic acid


Formula: C₁₀H₉BrO₂
MW: 241.08126 g/mol
MDL Number: MFCD29067870
CAS Number: 121146-16-5
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty