Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

N-Methylmorpholine-N-oxide 50% in aqueous solution

Supplier: APOLLO SCIENTIFIC
Synonyms: 4-Methylmorpholine 4-oxide4-Methylmorpholine N-Oxide

OR52933-250G OR52933-1KG
76591-448EA 79 USD InStock
76591-448 76591-446
N-Methylmorpholine-N-oxide 50% in aqueous solution
N-Methylmorpholine N-oxide


Formula: C₅H₁₁NO₂
MW: 117.15 g/mol
Boiling Pt: 119 °C
Density: 1.12
MDL Number: MFCD00005947
CAS Number: 7529-22-8
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty