Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Bromo-3-hydroxypyridine ≥95%

Supplier: APOLLO SCIENTIFIC
Synonyms: 2-Bromo-3-pyridinol

OR62149-100G OR62149-25G
76594-530EA 57 USD InStock
76594-530 76594-528
2-Bromo-3-hydroxypyridine ≥95%
2-Bromo-3-hydroxypyridine


Formula: C₅H₄BrNO
MW: 174 g/mol
MDL Number: MFCD00006220
CAS Number: 6602-32-0
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty