Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Magid® CutMaster® 1266KV Para-Aramid Lined Goatskin Leather Drivers Gloves, Cut Level A6
Supplier: Magid
Magid® CutMaster®
1266KVM 1266KVS 1266KVXXL 1266KVXL 1266KVL 1266KVXXXL
76635-708PR 27.81 USD InStock
76635-708 76635-718 76635-716 76635-710 76635-714 76635-712
Magid® CutMaster® 1266KV Para-Aramid Lined Goatskin Leather Drivers Gloves, Cut Level A6
Gloves
The 1266KV is a lightweight, comfortable goatskin leather driver gloves.
  • Goatskin leather shell provides dexterity and abrasion resistance
  • Stitched-in para-aramid blend knit fabric liner delivers an optimal fit and dependable cut protection
  • Gunn cut pattern increases comfort and dexterity
  • Keystone thumb delivers a premium, natural fit
  • Shirred elastic on back improves fit
  • ANSI Level A6 Cut Resistance, ANSI Level 3 Puncture Resistance
The 1266KV goatskin leather driver glove features a stitched-in para-aramid liner for added protection and a gunn cut thumb with a shirred elastic wrist for enhanced comfort.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty