Would you like to log in again?
M12 Ultra™ Electrophoresis and Visualization Package
Supplier: EDVOTEK
M12 Ultra™
505
470350-628EA 274.1 USD InStock
470350-628
M12 Ultra™ Electrophoresis and Visualization Package
Electrophoresis Systems Horizontal Gel Electrophoresis Systems
The M12 Ultra™ includes the M12 Complete™ Electrophoresis Package and the all-new TruBlu™ Jr. blue light transilluminator.
  • For 1 to 2 lab groups
  • Produce results in 15 to 30 minutes
  • Can be used to visualize of any blue-light reactive dyed agarose gels
  • Now able to examine green fluorescent protein (GFP)
This dynamic duo enables real-time visualization of SYBR® Safe or other blue light-reactive dye agarose gels.
For blue light-reactive gels, overlay the orange filter on the M12 lid and slide the TruBlu™ Jr. underneath for instant visualization!
The M12 Ultra™ offers the ultimate flexibility to either visualize gels in real-time with the orange contrast lid and TruBlu™ Jr., or to stain your gels post electrophoresis if you plan to use a traditional blue stain.
Bonus feature! The TruBlu™ Jr. can also be used to examine fluorescence from green fluorescent protein (GFP). This package supports one or two student groups in two standard length gel trays for experiments that require less separation, or one long gel tray for experiments that require more.
TruBlu™ Jr. Features:
• Battery operated for convenient use in the classroom
• Designed to work seamlessly with the EDVOTEK® M12 Electrophoresis System
• Includes an orange contrast lid
• Safe, non-UV wavelength
• Blue Light emission spectrum centered around 470 nm is compatible with SYBR® Safe, GelGreen® & other blue light-reactive dyes.
• 10×6 cm viewing surface
M12 Complete™ Features:
• Sleek new design speeds electrophoresis
• Includes complete set of new & improved electrophoresis accessories
• Now includes a DNA DuraGel™ (Cat. #S-43) for pipetting practice!
• Large color coded push tabs for easy lid insertion & removal
• Pour spout for buffer disposal
• Improved ventilation reduces lid condensation
• User replaceable electrodes
• Chamber reverse compatible with previous Edvotek® accessories
Packaging: This package includes: (2) 7×7 cm gel trays, (1) 14×7 cm gel tray, (2) 6/8 tooth combs, (4) rubber end caps, (1) DNA DuraGel™, (1) TruBlu™ Jr., (1) orange contrast lid.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty