Would you like to log in again?
Motic HDMI Microscope Camera
Supplier: NATIONAL OPTICAL & SCIENTIFIC SE
Motic
D-MOTICAM 1080X
470349-292EA 1189.9 USD InStock
470349-292
Motic HDMI Microscope Camera
Cameras
The Moticam 1080X is a camera that was designed for ease of use and the ability to output fast live images at 1920×1080 pixels in full HD.
Unlike other cameras, the Moticam 1080X does not require a computer. Simply plug the camera through the HDMI port and display images with the use of the onboard software and a USB mouse through an HDMI monitor.
By having an integrated SD card slot, not only does saving images not require a computer, but it also allows flexibility to save portable data.
Delivery information: Supplied with 12 V power adapter, CS ring, Motic 4-dot calibration slide, dust cap, manual, 12 mm lens, adapter ø30/ø38, macro tube, macro calibration card.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty