Would you like to log in again?
Motic 4K Microscope Camera
Motic
D-MOTICAM 4000
470349-294EA 1148.7 USD InStock
470349-294
Motic 4K Microscope Camera
Cameras
The Moticam 4000 is a camera that was designed for ease of use and the ability to output fast live images at 4K (3840×2160) pixels in full HD.
Unlike other cameras, the Moticam 4000 can be connected to a computer or through HDMI. Simply plug the camera through the HDMI port and display images with the use of the onboard software and a USB mouse through an HDMI monitor, or connect your camera through USB to a Windows, Mac or Linux computer.
By having an integrated SD card slot, not only does saving images not require a computer, but it also allows flexibility to save portable data.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty