Would you like to log in again?
Sterile Empty Vials, Assembled and Pyrogen Free
Supplier: ALK Life Science Science Solutions
SEV213S-AMB SEV20 SEV213-25-AMB SEV50-AMB SEV100 SEV1020G SEV520Y SEV520R SEV520G SEV520S SEV213 SEV213-25 SEV50-FLUR SEV520S-AMB SEV30 SEV1020R SEV100-FLUR SEV30-AMB SEV1020B SEV20-16 SEV1020Y SEV50 SEV1020S-FLUR SEV520B SEV1020S SEV30-FLUR SEV1020S-AMB SEV100-AMB
76640-552PK 431 USD InStock
76640-552 76640-574 76640-554 76640-576 76640-534 76640-556 76640-578 76640-558 76640-536 76994-118 76640-570 76640-572 76640-550 76994-122 76994-120 76640-564 76640-542 76640-544 76640-566 76640-568 76640-546 76640-548 76640-580 76640-582 76640-560 76640-540 76640-562 76640-538
Sterile Empty Vials, Assembled and Pyrogen Free
Vials
ALK Life Science Solutions provides high quality sterile empty vials to maintain the purity and integrity of what is inside.
  • Type 1 glass USP borosilicate
  • Latex free stopper
  • Produced at a cGMP FDA manufacturer
  • USP tested for sterility and endotoxins (pyrogen free)
  • Assembled in the USA, in an ISO 5 (class 100) cleanroom
Ideal for analytical laboratories for specimen containment and transfer. The sterilization processes for ALK Life Science solutions sterile empty vials are carried out under strict Class 100 U.S. Pharmacopoeia and Federal Code of Regulations methodologies. Assembled units are sterile and free of all endotoxins and meet the FDA's mandated 14-day sterility testing protocol. Meets all of the requirements of section 503B. Meet USP <1207>, <660>, <797>, <381>, <788>, <71>, and <85>.
Certifications: Certificate of analysis available for all lots.
Delivery information: Packaged and ready to use upon delivery.
Packaging: Tamper evident.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty