Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

(S)-2-Amino-3-cyclopentylpropanoic acid 97%

Supplier: Ambeed
Warning
Synonyms: H-Cpa-OH3-Cyclopentyl-L-alanine

A115914-250MG A115914-1G
76687-824EA 54.59 USD InStock
76687-824 76687-822
(S)-2-Amino-3-cyclopentylpropanoic acid 97%
(S)-2-Amino-3-cyclopentylpropanoic acid
Caution: for Research Use Only


Formula: C₈H₁₅NO₂
MW: 157.21 g/mol
MDL Number: MFCD01632069
CAS Number: 99295-82-6
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty