Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Boc-D-Tyr-OMe 96%

Supplier: Ambeed
Warning
Synonyms: N-Boc-D-tyrosine methyl ester

A115975-5G A115975-1G A115975-100G A115975-25G A115975-10G
76687-842EA 11.33 USD InStock
76687-842 76687-840 76687-838 76687-836 76687-834
Boc-D-Tyr-OMe 96%
Boc-D-Tyr-OMe
Caution: for Research Use Only


Formula: C₁₅H₂₁NO₅
MDL Number: MFCD00057824
CAS Number: 76757-90-9
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty