Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

D-(-)-Quinic acid 95%

Supplier: Ambeed
Warning
Synonyms: (1α,3R,4α,5R)-1,3,4,5-Tetrahydroxycyclohexanecarboxylic acid

A121034-1G A121034-100MG A121034-10G A121034-5G A121034-100G A121034-25G
76688-112EA 9.27 USD InStock
76688-112 76688-114 76688-110 76688-106 76688-116 76688-108
D-(-)-Quinic acid 95%
D(-)-Quinic acid
Caution: for Research Use Only


Formula: C₇H₁₂O₆
MW: 192.17 g/mol
MDL Number: MFCD00003864
CAS Number: 77-95-2
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty