Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Azelaic acid 98%

Supplier: Ambeed
Warning
Synonyms: 1,9-Nonanedioic acid1,7-Heptanedicarboxylic acid

A121847-100G A121847-500G A121847-25G
76688-156EA 33.99 USD InStock
76688-156 76688-152 76688-154
Azelaic acid 98%
Azelaic acid
Caution: for Research Use Only


Formula: C₉H₁₆O₄
MW: 188.22 g/mol
MDL Number: MFCD00004432
CAS Number: 123-99-9
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty