Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Benzylideneacetone 98%

Supplier: Ambeed
Warning
Synonyms: 4-Phenylbut-3-en-2-one4-Phenyl-3-buten-2-one

A155864-25G A155864-10G A155864-500G
76689-554EA 6.18 USD InStock
76689-554 76689-552 76689-556
Benzylideneacetone 98%
Benzylideneacetone
Caution: for Research Use Only


Formula: C₁₀H₁₀O
MW: 146.19 g/mol
MDL Number: MFCD00008779
CAS Number: 122-57-6
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty