Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Allantoin 98%

Supplier: Ambeed
Warning
Synonyms: 1-(2,5-Dioxoimidazolidin-4-yl)urea

A169387-100MG
76690-128EA 5.15 USD InStock
76690-128
Allantoin 98%
Allantoin
Caution: for Research Use Only


Formula: C₄H₆N₄O₃
MW: 158.12
MDL Number: MFCD00005260
CAS Number: 97-59-6
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty