Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

N-Carbamoyl-DL-aspartic acid 95%

Supplier: Ambeed
Warning
Synonyms: N-Carbamyl-DL-aspartic Acid2-Ureidosuccinic acid

A220289-1G
76691-790EA 5.15 USD InStock
76691-790
N-Carbamoyl-DL-aspartic acid 95%
N-Carbamoyl-DL-aspartic acid
Caution: for Research Use Only


Formula: C₅H₈N₂O₅
MW: 176.13 g/mol
MDL Number: MFCD00042822
CAS Number: 923-37-5
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty