Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Dimethyl-5-hydroxyisophthalate 98%

Supplier: Ambeed
Warning
Synonyms: 5-Hydroxyisophthalic acid dimethyl esterDimethyl 5-hydroxyisophthalate

A224614-100G A224614-25G A224614-10G A224614-500G
76691-904EA 8.24 USD InStock
76691-904 76691-902 76691-908 76691-906
Dimethyl-5-hydroxyisophthalate 98%
Dimethyl 5-hydroxyisophthalate
Caution: for Research Use Only


Formula: C₁₀H₁₀O₅
MW: 210.19 g/mol
MDL Number: MFCD00134367
CAS Number: 13036-02-7
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty