Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Diboron trioxide 99%

Supplier: Ambeed
Danger
Synonyms: Boron oxideBoric anhydride

A249794-25G
76692-448EA 6.18 USD InStock
76692-448
Diboron trioxide 99%
di-Boron trioxide
Caution: for Research Use Only


Formula: B₂O₃
MW: 69.62 g/mol
MDL Number: MFCD00011315
CAS Number: 1303-86-2
UN: 3077
ADR: 9,Ⅲ
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty