Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

α,α-Dimethoxytoluene 97%

Supplier: Ambeed
Warning
Synonyms: Benzaldehyde dimethyl acetal

A267373-25G A267373-500G A267373-100G A267373-1000G
77092-480EA 6.18 USD InStock
77092-480 76692-794 76692-792
α,α-Dimethoxytoluene 97%
α,α-Dimethoxytoluene
Caution: for Research Use Only


Formula: C₉H₁₂O₂
MW: 152.19 g/mol
MDL Number: MFCD00008491
CAS Number: 1125-88-8
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty