Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Nitro-o-anisic acid 98%

Supplier: Ambeed
Warning

A357473-25G A357473-1G A357473-5G
76694-162EA 15.45 USD InStock
76694-162 76694-166 76694-164
4-Nitro-o-anisic acid 98%
4-Nitro-o-anisic acid
Caution: for Research Use Only


Formula: C₈H₇NO₅
MW: 197.15 g/mol
MDL Number: MFCD00216573
CAS Number: 2597-56-0
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty