Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

8-Bromoadenine 95%

Supplier: Ambeed
Warning
Synonyms: 8-Bromo-1H-adenine8-Bromo-9H-purin-6-amine8-Bromo-7H-purin-6-amine

A385276-100MG A385276-250MG
76694-556EA 58.71 USD InStock
76694-556 76694-558
8-Bromoadenine 95%
8-Bromoadenine
Caution: for Research Use Only


Formula: C₅H₄BrN₅
MW: 214.02 g/mol
MDL Number: MFCD00082518
CAS Number: 6974-78-3
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty