Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

D-Homophenylalanine 98%

Supplier: Ambeed
Warning
Synonyms: H-D-HoPhe-OH(R)-2-Amino-4-phenylbutyric AcidH-D-HomoPhe-OH

A385892-10G A385892-100G A385892-1G
76694-566EA 263.68 USD InStock
76694-566 76694-568 76694-570
D-Homophenylalanine 98%
D-Homophenylalanine
Caution: for Research Use Only


Formula: C₁₀H₁₃NO₂
MW: 179.22 g/mol
MDL Number: MFCD00063091
CAS Number: 82795-51-5
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty