Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Diammonium tetrachloroplatinate 98%

Supplier: Ambeed
Danger
Synonyms: Ammonium tetrachloroplatinate(II)Ammonium tetrachloroplatinate

A389286-250MG
76694-616EA 29.87 USD InStock
76694-616
Diammonium tetrachloroplatinate 98%
Diammonium tetrachloroplatinate
Caution: for Research Use Only


Formula: H₈Cl₄N₂Pt
MW: 372.97 g/mol
MDL Number: MFCD00010885
CAS Number: 13820-41-2
UN: 3288
ADR: 6.1,Ⅲ
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty