Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Benzo[d]isoxazol-3-ol 98%

Supplier: Ambeed
Warning
Synonyms: 3-Hydroxy-1,2-benzisoxazole

A393339-250MG A393339-5G A393339-1G A393339-10G
76694-698EA 128.75 USD InStock
76694-698 76694-696 76694-694 76694-692
Benzo[d]isoxazol-3-ol 98%
Benzo[d]isoxazol-3-ol
Caution: for Research Use Only


Formula: C₇H₅NO₂
MW: 135.12 g/mol
MDL Number: MFCD00125030
CAS Number: 21725-69-9
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty