Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Bis(4-methoxyphenyl) disulfide 98%

Supplier: Ambeed
Danger
Synonyms: 1,2-Bis(4-methoxyphenyl)disulfane1,2-Bis(4-methoxyphenyl)disulphane

A476237-1G A476237-25G A476237-5G A476237-250MG
76695-454EA 13.39 USD InStock
76695-454 76695-456 76695-458 76695-460
Bis(4-methoxyphenyl) disulfide 98%
Bis(4-methoxyphenyl) disulphide
Caution: for Research Use Only


Formula: C₁₄H₁₄O₂S₂
MW: 278.4 g/mol
MDL Number: MFCD00041358
CAS Number: 5335-87-5
UN: 3077
ADR: 9,Ⅲ
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty