Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Chloroiodomethane 97% stabilized

Supplier: Ambeed
Warning

Stabilization: Stabilized with Copper chip

A482895-25G A482895-100G A482895-5G
76695-552EA 15.45 USD InStock
76695-552 76695-554 77551-466
Chloroiodomethane 97% stabilized
Chloroiodomethane
Caution: for Research Use Only


Formula: CH₂ClI
MW: 176.38 g/mol
MDL Number: MFCD00001078
CAS Number: 593-71-5
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty