Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Cyclohexyl ethyl ketone 99%

Supplier: Ambeed
Warning

A527235-250MG A527235-5G A527235-1G
76695-912EA 61.8 USD InStock
76695-912 76695-914 76695-916
Cyclohexyl ethyl ketone 99%
Cyclohexyl ethyl ketone
Caution: for Research Use Only


Formula: C₉H₁₆O
MDL Number: MFCD00060814
CAS Number: 1123-86-0
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty