Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3-Ethoxy-4-methoxybenzoic acid 95%

Supplier: Ambeed
Warning

A570216-1G
76696-292EA 46.35 USD InStock
76696-292
3-Ethoxy-4-methoxybenzoic acid 95%
3-Ethoxy-4-methoxybenzoic acid
Caution: for Research Use Only


Formula: C₁₀H₁₂O₃
MDL Number: MFCD00276591
CAS Number: 2651-55-0
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty