Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Copper(I) oxide 98%

Supplier: Ambeed
Danger
Synonyms: Cuprous oxideDicopper oxide

A575762-25G
76696-362EA 5.15 USD InStock
76696-362
Copper(I) oxide 98%
Copper (I) oxide
Caution: for Research Use Only


Formula: Cu₂O
MW: 143.09 g/mol
MDL Number: MFCD00010974
CAS Number: 1317-39-1
UN: 3077
ADR: 9,Ⅲ
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty