Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1,4-Butanediol dimethanesulfonate 98%

Supplier: Ambeed
Danger
Synonyms: Butane-1,4-diyl dimethanesulfonate1,4-Butanediol dimethanesulphonateButane-1,4-diyl dimethanesulphonate1,4-Butanediol dimethanesulfonate

A593389-1G A593389-1MG
76696-462EA 6.18 USD InStock
76696-462 76696-464
1,4-Butanediol dimethanesulfonate 98%
Busulfan
Caution: for Research Use Only


Formula: C₆H₁₄O₆S₂
MW: 246.29 g/mol
MDL Number: MFCD00007562
CAS Number: 55-98-1
UN: 2811
ADR: 6.1,Ⅲ
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty