Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

(±)-4-Chloromandelic acid 98%

Supplier: Ambeed
Warning
Synonyms: 4-Chloromandelic acid(±)-4-Chlorophenylglycolic acid

A705029-25G A705029-5G A705029-10G
76697-350EA 10.3 USD InStock
76697-350 76697-352 76697-354
(±)-4-Chloromandelic acid 98%
(±)-4-Chloromandelic acid
Caution: for Research Use Only


Formula: C₈H₇ClO₃
MW: 186.59 g/mol
MDL Number: MFCD00042724
CAS Number: 492-86-4
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty