Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Benzyl-N-boc-L-proline 95%

Supplier: Ambeed
Warning
Synonyms: (R)-2-Benzyl-1-(tert-butoxycarbonyl)pyrrolidine-2-carboxylic acid

A709836-100MG A709836-250MG
76697-392EA 112.27 USD InStock
76697-392 76697-390
2-Benzyl-N-boc-L-proline 95%
2-Benzyl-N-boc-L-proline
Caution: for Research Use Only


Formula: C₁₇H₂₃NO₄
MW: 305.37 g/mol
MDL Number: MFCD06795522
CAS Number: 706806-60-2
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty