Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Capric acid 98%

Supplier: Ambeed
Warning
Synonyms: Decanoic acidDecylic acid

A765305-100G A765305-25G A765305-500G
76697-786EA 9.27 USD InStock
76697-786 76697-782 76697-784
Capric acid 98%
Capric acid
Caution: for Research Use Only


Formula: C₁₀H₂₀O₂
MW: 172.27 g/mol
MDL Number: MFCD00004441
CAS Number: 334-48-5
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty