Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Benzoyl ferrocene 98%

Supplier: Ambeed
Warning
Synonyms: Benzoylferrocene

A765547-100G A765547-5G A765547-25G A765547-1G
76697-790EA 11.33 USD InStock
76697-790 76697-788 76697-792 76697-794
Benzoyl ferrocene 98%
Benzoylferrocene
Caution: for Research Use Only


Formula: C₁₇H₁₄FeO
MW: 290,14 g/mol
MDL Number: MFCD00001431
CAS Number: 1272-44-2
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty