Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Cyclobutyl methyl ketone 95%

Supplier: Ambeed
Danger
Synonyms: 1-Cyclobutylethan-1-one

A854722-5G A854722-1G A854722-250MG A854722-25G
76698-406EA 21.63 USD InStock
76698-406 76698-404 76698-408 76698-410
Cyclobutyl methyl ketone 95%
Cyclobutyl methyl ketone
Caution: for Research Use Only


Formula: C₆H₁₀O
MW: 98.15 g/mol
MDL Number: MFCD00037148
CAS Number: 3019-25-8
UN: 1224
ADR: 3,Ⅲ
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty