Would you like to log in again?
Synthware #35 Ball Stopper
Supplier: Kemtech America
Synthware
S450035
60004-858EA 38.3 USD InStock
60004-858
Synthware #35 Ball Stopper
Stoppers Glass Stoppers
35/20 ball joint penny head stopper.
  • Heavy wall construction
  • 33 expansion borosilicate glass
  • Standard ground glass joints work with other brands
For use with 35/20 socket joint.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty