Would you like to log in again?
Atomic Spectra Optical Bench
Supplier: Shiv Dial Sud & Sons
ASE1-C
470331-756EA 896.85 USD InStock
470331-756
Atomic Spectra Optical Bench
Educational Materials Physics Educational Materials Optics Learning Activities
High precision, quantitative atomic spectra experiment.
  • Optical bench provides precise optical alignment
  • Rotating table with mounted diffraction grating and telescope
  • Built-in power supply and spectrum tubes included
The optical alignment of the components is attained by optical bench, making set-up faster and experimentation easy. The telescope on the rotating table is mounted with bearings for smooth rotation. The diffraction grating is mounted at the center of the rotating table. The telescope is easy to focus and operate.
The rotating arm is graduated for accurate measurement of the spectral lines. The built-in power supply makes a more compact system with easier changing of the discharge tubes. Connection to power is easy, and there are no unnecessary wire intrusions.
Ordering information: Kit includes: Optical bench set (0.8 m), converging lens holder, adjustable collimating slit holder, spectrum tube power supply assembly, telescope assembly, and five spectrum tubes (hydrogen, helium, mercury, krypton, argon).
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty