Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Steel Burette Double Clamp
Supplier: EISCO SCIENTIFIC LLC
CH0254B
470344-078EA 7.79 USD InStock
470344-078
Steel Burette Double Clamp
Clamps General Purpose Clamps
Steel burette double clamp featuring a 7" separation and a maximum rod diameter of 20 mm.
  • Steel burette double clamp, capable of holding one or two burettes simultaneously
  • Rubber coated clamp jaws keep burettes safe but secure
  • Clamp holds burettes approximately 7 inches apart
  • Steel clamp is durable and lightweight
  • Good for use in side-by-side comparison experiments, such as comparative titration
Double-sided steel clamp designed for the safe handling of burettes. The clamp tips are rubber-coated to protect equipment. Hold one or two burettes simultaneously. Adjustable clamp can be attached to rod stand and can hold burettes up to approximately 1" (approx. 3 cm). This double clamp is perfect for side-by-side experiments and demonstrations.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty